DMOG ≥97.0% (by GC)

Supplier: TCI America

Synonyms: N-[Methoxy(oxo)acetyl]glycine methyll ester, Dimethyloxaloylglycine, Methyl 2-((2-methoxy-2-oxoethyl)amino)-2-oxoacetate, DMOG

D5480-50MG D5480-250MG
TCD5480-50MGEA 51 USD
TCD5480-50MG TCD5480-250MG
DMOG ≥97.0% (by GC)
DMOG
Formula: C₆H₉NO₅
MW: 175.14 g/mol
Boiling Pt: 182 °C
MDL Number: MFCD05865098
CAS Number: 89464-63-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 49.0 to 53.0 deg-C
Acid Value max. 10.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR