Tyrphostin A9 ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: [4-Hydroxy-3,5-bis(2-methyl-2-propanyl)benzylidene]malononitrile, Malonoben

T3667-100MG T3667-25MG
TCT3667-100MGEA 207 USD
TCT3667-100MG TCT3667-25MG
Tyrphostin A9 ≥98.0% (by GC)
Tyrphostin A9
Formula: C₁₈H₂₂N₂O
MW: 282.387 g/mol
MDL Number: MFCD00209853
CAS Number: 10537-47-0
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 139.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR