6,7-Dihydrobenzo[b]thiophen-4-one ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
4-Oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-Dihydro-4-benzo[b]thiophenone, 4-Keto-4,5,6,7-tetrahydrothianaphthene
D5714-1G
D5714-5G
TCD5714-1GEA
39
USD
TCD5714-1G
TCD5714-5G
6,7-Dihydrobenzo[b]thiophen-4-one ≥95.0% (by GC)
6,7-Dihydrobenzo[b]thiophen-4-one
Formula:
C₈H₈OS MW: 152.211 g/mol Boiling Pt: 92 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005861 CAS Number: 13414-95-4 EINECS: 236-510-7 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to lump |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...