2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
DL-2,3-Dimercapto-1-propanesulphonic acid, sodium salt monohydrate, DMPS, Sodium 2,3-dimercaptopropane-1-sulphonate hydrate, 2,3-Dimercaptopropanesulphonic acid sodium salt, Sodium 2,3-Dimercaptopropanesulphonate monohydrate
S0524-25G
S0524-5G
TCS0524-25GEA
394
USD
TCS0524-25G
TCS0524-5G
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate ≥97.0% (by titrimetric analysis)
2,3-Dimercapto-1-propanesulfonic acids odium salt monohydrate
Formula:
C₃H₉NaO₄S₃·H₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00149543 CAS Number: 207233-91-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % (calcd.on anh.substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...