1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene, 1-Ethoxy-4-(p-tolylethynyl)benzene

E1342-1G E1342-5G
TCE1342-1GEA 63 USD
TCE1342-1G TCE1342-5G
1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene ≥98.0% (by GC)
1-Ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene
Formula: C₁₇H₁₆O
MDL Number: MFCD01817758
CAS Number: 116903-46-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR