Methacrylic anhydride ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Methacrylic Anhydride [stabilized with 1,1,3-Tris(3-tert-butyl-4-hydroxy-6-methylphenyl)butane]

M3139-100G M3139-500G
TCM3139-100GEA 52 USD
TCM3139-100G TCM3139-500G
Methacrylic anhydride ≥97.0% (by GC, titration analysis)
Methacrylic anhydride
Formula: C₈H₁₀O₃
MW: 154.165 g/mol
Boiling Pt: 205 °C
Flash Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008586
CAS Number: 760-93-0
EINECS: 212-084-8

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Morpholine Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR