Propyl vinyl ether ≥97.0% (by GC) stabilized

Supplier: TCI America
Danger


Stabilization: Stabilised with potassium hydroxide

P2424-250ML P2424-25ML
TCP2424-250MLEA 76 USD
TCP2424-250ML TCP2424-25ML
Propyl vinyl ether ≥97.0% (by GC) stabilized
Propyl vinyl ether
Formula: C₅H₁₀O
Boiling Pt: 65 °C
Flash Pt: −26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080707
CAS Number: 764-47-6
EINECS: 212-123-9
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR