2-(Perfluorobutyl)ethyl acrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

N1107-5G N1107-25G
TCN1107-5GEA 49 USD
TCN1107-5G TCN1107-25G
2-(Perfluorobutyl)ethyl acrylate ≥98.0% (by GC) stabilized
2-(Perfluorobutyl)ethyl acrylate
Formula: C₉H₇F₉O₂
MDL Number: MFCD00236104
CAS Number: 52591-27-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR