2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

F1122-1G F1122-5G
TCF1122-1GEA 53 USD
TCF1122-1G TCF1122-5G
2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl
Formula: C₃₂H₄₅F
MDL Number: MFCD11053473
CAS Number: 106349-49-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 105 to 280 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR