trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(trans-4-Pentylcyclohexyl)-4-ethoxybenzene, 1-Ethoxy-4-(trans-4-amylcyclohexyl)benzene, 1-Ethoxy-4-(trans-4-pentylcyclohexyl)benzene

E1367-5G E1367-25G
TCE1367-5GEA 40 USD
TCE1367-5G TCE1367-25G
trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene ≥98.0% (by GC)
trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene
Formula: C₁₉H₃₀O
MW: 274.448 g/mol
MDL Number: MFCD06658196
CAS Number: 84540-32-9
EINECS: 283-125-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR