trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate ≥98.0% (by GC)

Supplier: TCI America

P2696-1G P2696-5G
TCP2696-1GEA 66 USD
TCP2696-1G TCP2696-5G
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate
Formula: C₃₃H₅₂O₂
MDL Number: MFCD11053459
CAS Number: 131790-57-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 211.0 to 237.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR