Trifluoroacetaldehyde hydrate (contains Total ca. 35% Water)

Supplier: TCI America
Danger

Synonyms: Trifluoroacetaldehyde hydrate, 2,2,2-Trifluoro-1,1-ethanediol

T3654-25G T3654-5G
TCT3654-25GEA 88 USD
TCT3654-25G TCT3654-5G
Trifluoroacetaldehyde hydrate (contains Total ca. 35% Water)
Trifluoroacetaldehyde hydrate
Formula: C₂H₃F₃O₂
MW: 116.039 g/mol
MDL Number: MFCD00167359
CAS Number: 421-53-4
UN: 1989
ADR: 3 II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Water 32.0 to 37.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR