Iron(III) meso-tetraphenylporphine chloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

I0937-1G I0937-200MG
TCI0937-1GEA 170 USD
TCI0937-1G TCI0937-200MG
Iron(III) meso-tetraphenylporphine chloride ≥97.0% (by HPLC, titration analysis)
Iron(III) meso-tetraphenylporphine chloride
Formula: C₄₄H₂₈ClFeN₄
Melting Pt: 300 °C
MDL Number: MFCD00012152
CAS Number: 16456-81-8

Order Now

Specification Test Results

Appearance Green to Dark green to Dark blue powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Chelometric Titration) min. 97.0 %
Molar extinction coefficient min. 100000 (THF, 414 to 418nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR