trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Ethoxy-4-(trans-4-propylcyclohexyl)benzene

E1369-25G E1369-5G
TCE1369-25GEA 167 USD
TCE1369-25G TCE1369-5G
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene ≥98.0% (by GC)
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene
Formula: C₁₇H₂₆O
MW: 246.394 g/mol
MDL Number: MFCD06658195
CAS Number: 80944-44-1
EINECS: 279-633-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR