RITA ≥98.0% (by GC)

Supplier: TCI America

F1235-25MG F1235-5MG
TCF1235-25MGEA 374 USD
TCF1235-25MG TCF1235-5MG
RITA ≥98.0% (by GC)
RITA
Formula: C₁₄H₁₂O₃S₂
Storage Temperature: Refrigerator
MDL Number: MFCD03235294
CAS Number: 213261-59-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 158.0 to 162.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR