Methoxyacetic anhydride ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Methoxyacetic Anhydride

M3180-25G M3180-5G
TCM3180-25GEA 138 USD
TCM3180-25G TCM3180-5G
Methoxyacetic anhydride ≥96.0% (by GC, titration analysis)
Methoxyacetic anhydride
Formula: C₆H₁₀O₅
MW: 162.141 g/mol
Boiling Pt: 120 °C
MDL Number: MFCD11976032
CAS Number: 19500-95-9
EINECS: 243-114-8

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR