Nefopam hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 5-Methyl-1-phenyl-3,4,5,6-tetrahydro-1H-benzo[f][1,4]oxazocine hydrochloride

N1169-1G N1169-5G
TCN1169-1GEA 41 USD
TCN1169-1G TCN1169-5G
Nefopam hydrochloride ≥98.0% (by HPLC, titration analysis)
Nefopam hydrochloride
Formula: C₁₇H₂₀ClNO
Melting Pt: 251 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012750
CAS Number: 23327-57-3
EINECS: 245-585-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,06440

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR