Tazobactam ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

T3732-1G T3732-5G
TCT3732-1GEA 72 USD
TCT3732-1G TCT3732-5G
Tazobactam ≥97.0% (by HPLC, titration analysis)
Tazobactam
Formula: C₁₀H₁₂N₄O₅S
MDL Number: MFCD00867002
CAS Number: 89786-04-9
Merck Index: 14,09083

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +162 to +167 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR