Mitoxantrone dihydrochloride ≥97.0% (by HPLC)
Supplier: TCI America
|
Danger
|
Synonyms:
Novantrone, 1,4-Dihydroxy-5,8-bis((2-((2-hydroxyethyl)amino)ethyl)amino)anthracene-9,10-dione dihydrochloride
M3110-100MG
M3110-25MG
TCM3110-100MGEA
102
USD
TCM3110-100MG
TCM3110-25MG
Mitoxantrone dihydrochloride ≥97.0% (by HPLC)
Mitoxantrone dihydrochloride
Formula:
C₂₂H₃₀Cl₂N₄O₆ MW: 517.404 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00242943 CAS Number: 70476-82-3 EINECS: 274-619-1 UN: 2811 ADR: 6.1,II Merck Index: 14,06217 |
Specification Test Results
Appearance | Dark green to Dark purple to Black powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...