Trimethylboroxine ∼50% in THF

Supplier: TCI America
Danger

T3810-25G T3810-5G
TCT3810-25GEA 144 USD
TCT3810-25G TCT3810-5G
Trimethylboroxine ∼50% in THF
Trimethylboroxine
Formula: C₃H₉B₃O₃
Flash Pt: −20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013354
CAS Number: 823-96-1
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (Neutralization titration) 45.0 to 55.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR