Formic acid-triethylamine 5:2 addition compound ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Formic acid triethylamine complex 5:2, Formic acid - triethylamine (5:2) Azeotrope, TEAF

F1202-100ML F1202-25ML
TCF1202-100MLEA 141 USD
TCF1202-100ML TCF1202-25ML
Formic acid-triethylamine 5:2 addition compound ≥98.0% (by titrimetric analysis)
Formic acid-triethylamine 5:2 addition compound
Formula: C₁₇H₄₀N₂O₁₀
MW: 432.51 g/mol
MDL Number: MFCD00040600
CAS Number: 15077-13-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR