4-Methoxy-6-methyl-2H-pyran-2-one ≥98.0% (by GC)

Supplier: TCI America

M2881-1G M2881-200MG
TCM2881-1GEA 96 USD
TCM2881-1G TCM2881-200MG
4-Methoxy-6-methyl-2H-pyran-2-one ≥98.0% (by GC)
4-Methoxy-6-methyl-2H-pyran-2-one
Formula: C₇H₈O₃
MDL Number: MFCD00006640
CAS Number: 672-89-9
EINECS: 211-600-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR