(-)-Epicatechin ≥97.0% (by HPLC)

Supplier: TCI America
Warning

E1226-1G E1226-5G
TCE1226-1GEA 130 USD
TCE1226-1G TCE1226-5G
(-)-Epicatechin ≥97.0% (by HPLC)
(-)-Epicatechin
Formula: C₁₅H₁₄O₆
Melting Pt: 240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075648
CAS Number: 490-46-0
EINECS: 207-710-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D -56.0 to -64.0 deg (C=0.1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR