Fmoc-Chg-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Fmoc-L-Chg-OH, Fmoc-cyclohexyl-Gly-OH, N-Fmoc-L-cyclohexylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-cyclohexylglycine, N-Fmoc-2-cyclohexyl-L-glycine

F1248-1G F1248-5G
TCF1248-1GEA 35 USD
TCF1248-1G TCF1248-5G
Fmoc-Chg-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Chg-OH
Formula: C₂₃H₂₅NO₄
MW: 379.456 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00190878
CAS Number: 161321-36-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 181.0 to 185.0 deg-C
Specific rotation [a]20/D -3.0 to -140.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR