Fmoc-Tle-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F1251-25G F1251-5G
TCF1251-25GEA 213 USD
TCF1251-25G TCF1251-5G
Fmoc-Tle-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Tle-OH
Formula: C₂₁H₂₃NO₄
MDL Number: MFCD00065649
CAS Number: 132684-60-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR