Fmoc-N-Me-Val-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-valine, Fmoc-N-Me-Val-OH, N-Fmoc-N-methyl-L-valine
F1165-1G
F1165-5G
TCF1165-1GEA
53
USD
TCF1165-1G
TCF1165-5G
Fmoc-N-Me-Val-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-N-Me-Val-OH
Formula:
C₂₁H₂₃NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00153395 CAS Number: 84000-11-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 188.0 to 192.0 deg-C |
Specific rotation [a]20/D | -68.0 to -72.0 deg (C=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...