Polyethylene glycol dimethacrylate n=∼14 stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2760-25G P2760-500G
TCP2760-25GEA 46.11 USD
TCP2760-25G TCP2760-500G
Polyethylene glycol dimethacrylate n=∼14 stabilized
Polyethylene glycol dimethacrylate
Formula: (C₂H₄O)nC₈H₁₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00014932
CAS Number: 25852-47-5

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
The average value of n 13.0 to 15.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR