Polyethylene glycol dimethacrylate n=∼9 stabilised

Supplier: TCI America
Warning

Synonyms: PEGDMA, Polyethylene Glycol Dimethacrylate average Mn 550, PEG dimethacrylate, Polyethylene Glycol Dimethacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2709-25G P2709-500G
TCP2709-25GEA 26.06 USD
TCP2709-25G TCP2709-500G
Polyethylene glycol dimethacrylate n=∼9 stabilised
Polyethylene glycol dimethacrylate
Formula: (C₂H₄O)nC₈H₁₀O₃
MDL Number: MFCD00014932
CAS Number: 25852-47-5

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
The average value of n 8.5 to 9.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR