Esculetin ≥99.0% (by HPLC, titration analysis) matrix for MALDI-TOF/MS

Supplier: TCI America
Warning

Synonyms: 6,7-Dihydroxycoumarin, 6,7-Dihydroxy-2H-chromen-2-one

E1287-1G E1287-200MG
TCE1287-1GEA 98 USD
TCE1287-1G TCE1287-200MG
Esculetin ≥99.0% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
Esculetin
Formula: C₉H₆O₄
MW: 178.14 g/mol
Melting Pt: 273 °C
MDL Number: MFCD00006874
CAS Number: 305-01-1
EINECS: 206-161-5
Merck Index: 14,03697

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR