1,3-Dibromoadamantane ≥99.0% (by GC), purified by sublimation

Supplier: TCI America

D5654-1G D5654-5G
TCD5654-1GEA 78 USD
TCD5654-1G TCD5654-5G
1,3-Dibromoadamantane ≥99.0% (by GC), purified by sublimation
1,3-Dibromoadamantane
Formula: C₁₀H₁₄Br₂
MW: 294.03 g/mol
MDL Number: MFCD00077218
CAS Number: 876-53-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR