[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥99.5% (by GC), purified by sublimation
Supplier: TCI America
|
Warning
|
Synonyms:
Pyrromethene 546, 5,5-Difluoro-1,3,7,9,10-pentamethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide, BODIPY 493/503
D5724-1G
D5724-200MG
TCD5724-1GEA
210
USD
TCD5724-1G
TCD5724-200MG
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥99.5% (by GC), purified by sublimation
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane)
Formula:
C₁₄H₁₇BF₂N₂ MW: 262.11 g/mol Melting Pt: 257 °C |
MDL Number:
MFCD00467372 CAS Number: 121207-31-6 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 99.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...