Methyl-2-bromo-4-chlorobenzoate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromo-4-chlorobenzoic acid methyl ester, Methyl 2-bromo-4-chlorobenzoate
M3131-25G
M3131-5G
TCM3131-25GEA
201
USD
TCM3131-25G
TCM3131-5G
Methyl-2-bromo-4-chlorobenzoate ≥98.0% (by GC)
Methyl-2-bromo-4-chlorobenzoate
Formula:
C₈H₆BrClO₂ MW: 249.4900 g/mol |
MDL Number:
MFCD10566822 CAS Number: 57381-62-1 |
Specification Test Results
Appearance | Colourless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...