Ethyl-(S)-2-amino-4-phenylbutanoate hydrochloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: L-Homophenylalanine ethyl ester hydrochloride, H-HomoPhe-OEt·HCl, H-HoPhe-OEt.HCl, H-HoPhe-OEt·HCl

H1532-1G H1532-5G
TCH1532-1GEA 28 USD
TCH1532-1G TCH1532-5G
Ethyl-(S)-2-amino-4-phenylbutanoate hydrochloride ≥97.0% (by HPLC, titration analysis)
Ethyl-(S)-2-amino-4-phenylbutanoate hydrochloride
Formula: C₁₂H₁₇NO₂·HCl
MW: 243.7 g/mol
MDL Number: MFCD00190691
CAS Number: 90891-21-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 158.0 to 162.0 deg-C
Specific rotation [a]20/D +37.0 to +40.0 deg (C=1,H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR