4-Ethynyl-1,1'-biphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Ethynylbiphenyl, 4-Biphenylacetylene

E1141-1G E1141-5G
TCE1141-1GEA 53 USD
TCE1141-1G TCE1141-5G
4-Ethynyl-1,1'-biphenyl ≥98.0% (by GC)
4-Ethynyl-1,1'-biphenyl
Formula: C₁₄H₁₀
MW: 178.2292 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00102191
CAS Number: 29079-00-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR