1-Methyl-1H-pyrazol-5-ol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Methyl-1H-pyrazol-5-ol, 5-Hydroxy-1-methyl-1H-pyrazole

H1695-1G H1695-5G
TCH1695-1GEA 66 USD
TCH1695-1G TCH1695-5G
1-Methyl-1H-pyrazol-5-ol ≥98.0% (by GC)
1-Methyl-1H-pyrazol-5-ol
Formula: C₄H₆N₂O
MW: 98.1032 g/mol
MDL Number: MFCD05864418
CAS Number: 33641-15-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR