(3-Fluoro-4-hydroxyphenyl)boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-Fluoro-4-hydroxyphenylboronic acid, 3-Fluoro-4-hydroxybenzeneboronic acid

F1132-1G F1132-5G
TCF1132-1GEA 93 USD
TCF1132-1G TCF1132-5G
(3-Fluoro-4-hydroxyphenyl)boronic acid (contains varying amounts of Anhydride)
(3-Fluoro-4-hydroxyphenyl)boronic acid
Formula: C₅H₅BFNO₂
MDL Number: MFCD06659838
CAS Number: 182344-14-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR