6-Hydroxy-7-methoxyquinazolin-4(1H)-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-Hydroxy-7-methoxy-4(3H)-quinazolinone, 6-Hydroxy-7-methoxy-3H-quinazolin-4-one

H1606-5G H1606-1G
TCH1606-5GEA 201 USD
TCH1606-5G TCH1606-1G
6-Hydroxy-7-methoxyquinazolin-4(1H)-one ≥98.0% (by HPLC, titration analysis)
6-Hydroxy-7-methoxyquinazolin-4(1H)-one
Formula: C₉H₈N₂O₃
MW: 192.1714 g/mol
MDL Number: MFCD20547768
CAS Number: 179688-52-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR