2-(1H-Imidazol-1-yl)acetonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-(Cyanomethyl)imidazole, (1-Imidazolyl)acetonitrile

I0990-1G I0990-5G
TCI0990-1GEA 141 USD
TCI0990-1G TCI0990-5G
2-(1H-Imidazol-1-yl)acetonitrile ≥98.0% (by GC, titration analysis)
2-(1H-Imidazol-1-yl)acetonitrile
Formula: C₅H₅N₃
MW: 107.12 g/mol
MDL Number: MFCD06421433
CAS Number: 98873-55-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR