3,3-Difluoropiperidine hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D5573-1G D5573-200MG
TCD5573-1GEA 163 USD
TCD5573-1G TCD5573-200MG
3,3-Difluoropiperidine hydrochloride ≥97.0% (by titrimetric analysis)
3,3-Difluoropiperidine hydrochloride
Formula: C5H10ClF2N
MDL Number: MFCD03452800
CAS Number: 496807-97-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR