2-Ethyl-1H-benzo[d]imidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E1358-25G E1358-5G
TCE1358-25GEA 258 USD
TCE1358-25G TCE1358-5G
2-Ethyl-1H-benzo[d]imidazole ≥98.0% (by GC, titration analysis)
2-Ethyl-1H-benzo[d]imidazole
Formula: C₉H₁₀N₂
MDL Number: MFCD00022684
CAS Number: 1848-84-6
EINECS: 217-433-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR