2,4-Dichloro-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D5580-25G D5580-5G
TCD5580-25GEA 111 USD
TCD5580-25G TCD5580-5G
2,4-Dichloro-3-nitropyridine ≥98.0% (by GC)
2,4-Dichloro-3-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
Boiling Pt: 78 °C
MDL Number: MFCD09261088
CAS Number: 5975-12-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR