(R)-(+)-3-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: (R)-3-Piperidinol hydrochloride, (3R)-piperidin-3-ol hydrochloride, (R)-Piperidin-3-ol hydrochloride, (R)-3-Hydroxypiperidine hydrochloride

H1537-1G H1537-5G
TCH1537-1GEA 24 USD
TCH1537-1G TCH1537-5G
(R)-(+)-3-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)
(R)-(+)-3-Hydroxypiperidine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.607 g/mol
MDL Number: MFCD00192221
CAS Number: 198976-43-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 195.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR