N-Methyl-3-pyrrolidinol ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Hydroxy-1-methylpyrrolidine, 1-Methyl-3-pyrrolidinol

M2115-25G M2115-5G
TCM2115-25GEA 348 USD
TCM2115-25G TCM2115-5G
N-Methyl-3-pyrrolidinol ≥97.0% (by titrimetric analysis)
N-Methyl-3-pyrrolidinol
Formula: C₅H₁₁NO
Boiling Pt: 75 °C
Flash Pt: 71 °C
MDL Number: MFCD00003176
CAS Number: 13220-33-2
EINECS: 236-189-3

Order Now

Specification Test Results

Appearance Colourless to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR