(S)-3-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: (S)-3-Piperidinol hydrochloride

H1538-5G H1538-1G
TCH1538-5GEA 151 USD
TCH1538-5G TCH1538-1G
(S)-3-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)
(S)-3-Hydroxypiperidine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.6100 g/mol
MDL Number: MFCD03426348
CAS Number: 475058-41-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 195.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR