4-Fluoro-2-methoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

F1003-1G F1003-5G
TCF1003-1GEA 96 USD
TCF1003-1G TCF1003-5G
4-Fluoro-2-methoxyaniline ≥98.0% (by GC, titration analysis)
4-Fluoro-2-methoxyaniline
Formula: C₇H₈FNO
Boiling Pt: 215 °C
MDL Number: MFCD00077536
CAS Number: 450-91-9
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR