4-Perhydroazepinone hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 4-Azepanone hydrochloride, Hexahydro-4-azepinone hydrochloride

H1673-1G H1673-5G
TCH1673-1GEA 26 USD
TCH1673-1G TCH1673-5G
4-Perhydroazepinone hydrochloride ≥98.0% (by titrimetric analysis)
4-Perhydroazepinone hydrochloride
Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
MDL Number: MFCD02183487
CAS Number: 50492-22-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR