8-Hydroxyquinoline-2-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

H1713-1G H1713-200MG
TCH1713-1GEA 95 USD
TCH1713-1G TCH1713-200MG
8-Hydroxyquinoline-2-carbaldehyde ≥98.0% (by GC)
8-Hydroxyquinoline-2-carbaldehyde
Formula: C₁₀H₇NO₂
MDL Number: MFCD00168962
CAS Number: 14510-06-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C
Acid Value max. 5.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR