4-(Pyrrolidin-1-yl)benzonitrile ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Danger

P2428-1G P2428-5G
TCP2428-1GEA 75 USD
TCP2428-1G TCP2428-5G
4-(Pyrrolidin-1-yl)benzonitrile ≥96.0% (by GC, titration analysis)
4-(Pyrrolidin-1-yl)benzonitrile
Formula: C₁₁H₁₂N₂
MDL Number: MFCD07368512
CAS Number: 10282-30-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 81.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR