Heparin sodium salt (from hog intestine)

Supplier: TCI America

Synonyms: Reviparin sodium

H0393-100MG H0393-1G
TCH0393-100MGEA 46.09 USD
TCH0393-100MG TCH0393-1G
Heparin sodium salt (from hog intestine)
Heparin sodium salt
MDL Number: MFCD00081689
CAS Number: 9041-08-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Total Nitrogen 1.3 to 2.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR