2,5-Dimethoxyphenol ≥98.0% (by GC)

Supplier: TCI America
Warning

D5666-1G D5666-200MG
TCD5666-1GEA 240 USD
TCD5666-1G TCD5666-200MG
2,5-Dimethoxyphenol ≥98.0% (by GC)
2,5-Dimethoxyphenol
Formula: C₈H₁₀O₃
Boiling Pt: 135 °C
MDL Number: MFCD00094237
CAS Number: 18113-18-3

Order Now

Specification Test Results

Appearance Colourless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR